Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC STK11 Human His-SU 5ug
SDP

Catalog No. 5000196375
Encompass_Preferred

Recombinant human STK11 (LKB1) kinase, expressed in E. coli with an N-terminal SUMO and 6xHis tag, supplied lyophilized for research applications. The protein regulates AMPK-family kinases and other substrates, and is provided for biochemical assays, kinase activity studies, and antibody or assay development.

  • Recombinant human source (430 amino acids).
  • N-terminal SUMO and 6xHis tag to facilitate purification.
  • High purity greater than 90% by reducing SDS-PAGE.
  • Available in multiple package sizes, including small research quantities.
  • Lyophilized formulation for improved stability and handling.
  • Reconstitution recommended at or above 100 μg/mL in ddH2O.
  • Store frozen; aliquot and keep at -20°C or -80°C for extended storage.

Catalog No. 50-001-96375 Supplier Medchemexpress LLC Supplier No. HYP715495UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human STK11 (LKB1) kinase, expressed in E. coli with an N-terminal SUMO and 6xHis tag, supplied lyophilized for research applications. The protein regulates AMPK-family kinases and other substrates, and is provided for biochemical assays, kinase activity studies, and antibody or assay development.

  • Recombinant human source (430 amino acids).
  • N-terminal SUMO and 6xHis tag to facilitate purification.
  • High purity greater than 90% by reducing SDS-PAGE.
  • Available in multiple package sizes, including small research quantities.
  • Lyophilized formulation for improved stability and handling.
  • Reconstitution recommended at or above 100 μg/mL in ddH2O.
  • Store frozen; aliquot and keep at -20°C or -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.