Learn More
Aviva Systems Biology Corp STK16 Antibody - middle region (ARP48229_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48229_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STK16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human STK16. Alternate names: FLJ39635, KRCT, MPSK, PKL12, TSF1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59768