Learn More
Medchemexpress LLC HY-101270 5mg Medchemexpress, STK16-IN-1 CAS:1223001-53-3 Purity:>98%
SDP
Catalog No. 501932080
Encompass_Preferred
Medchemexpress, HY-101270 5mg STK16-IN-1 CAS:1223001-53-3 STK16-IN-1 is a STK16 kinase inhibitor with an IC 50 of 295 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2080 Supplier Medchemexpress LLC Supplier No. HY1012705MG
Medchemexpress, HY-101270 5mg STK16-IN-1 CAS:1223001-53-3 STK16-IN-1 is a STK16 kinase inhibitor with an IC 50 of 295 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.