Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human STK16 (PKL12), His Tag Recombinant Protein

Catalog No. PR7859A
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR7859A 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR7859A Supplier Invitrogen™ Supplier No. PR7859A
Add to Cart
Edge
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

STK16 (PKL12) is a serine/threonine kinase that functions in signal transduction pathways and transcriptional regulation. Protein kinase that act on both serine and threonine residues. STK16 may regulate transforming growth factor-beta (Tgfb) and vascular endothelial growth factor transcription.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75716
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 8576
Molecular Weight (g/mol) 38.7 kDa
Name Human STK16 (PKL12), His Tag
Quantity 100 μg
Gene Alias Edpk; Embryo-derived protein kinase; F52; hPSK; kinase related to cerevisiae and thaliana; Krct; MPSK; Mpsk1; Myristoylated and palmitoylated serine/threonine-protein kinase; PKL12; protein kinase expressed in day 12 fetal liver; Protein kinase Krct; Protein kinase PKL12; serine/threonine kinase 16; serine/threonine-protein kinase 16; Stk16; TGF-beta-stimulated factor 1; Tsf1; TSF-1; tyrosine-protein kinase STK16
Common Name STK16
Kinase Family NAK Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.