Learn More
Medchemexpress LLC STK17A/B-IN-1 hydrochloride | 490.00 | 1 ML
SDP
Catalog No. 5000394101
Encompass_Preferred
STK17A/B-IN-1 hydrochloride is an orally active, potent, and selective STK17A/B inhibitor with an IC50 value of 23 nM for STK17A. It can be used in tumor research.
  • Orally active
  • Potent and selective STK17A/B inhibitor
  • IC50 value of 23 nM for STK17A
  • Can be used in tumor research
Catalog No. 50-003-94101 Supplier Medchemexpress LLC Supplier No. HY163626A10MM/1ML
STK17A/B-IN-1 hydrochloride is an orally active, potent, and selective STK17A/B inhibitor with an IC50 value of 23 nM for STK17A. It can be used in tumor research.
  • Orally active
  • Potent and selective STK17A/B inhibitor
  • IC50 value of 23 nM for STK17A
  • Can be used in tumor research