Learn More
Aviva Systems Biology Corp STK19 Antibody - middle region (ARP65510_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65510_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STK19. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human STK19. Alternate names: D6S60, D6S60E, G11, HLA-RP1, MGC117388, RP1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-288-511