Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human STK22B (TSSK2), His Tag Recombinant Protein

Catalog No. PV3622
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3622 10 μg
PR5947A 100 μg
PV4062 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV3622 Supplier Invitrogen™ Supplier No. PV3622
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

STK22B (TSSK2) is a serine/threonine kinase essential for the function and production of sperm (spermiogenesis). Deletions in the STK22B coding region correlate with numerous cardiovascular and glandular malformations. TSSK2 gene ontology annotations related to this gene include intracellular signal transduction; peptidyl-serine phosphorylation; protein autophosphorylation; protein phosphorylation; spermatid development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96PF2
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol, 150mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 23617
Molecular Weight (g/mol) 45.1 kDa
Name Human STK22B (TSSK2), His Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias DGSG; DGS-G; DiGeorge syndrome protein G; serine/threonine kinase 22B (spermiogenesis associated); serine/threonine-protein kinase 22B; spermiogenesis associated 2; SPOGA2; STK22B; testicular secretory protein Li 61; testis specific serine kinase 2; testis specific serine threonine kinase 2; testis specific serine/ threonine kinase 2; testis-specific kinase 2; testis-specific serine kinase 2; testis-specific serine/threonine-protein kinase 2; TSK2; TSK-2; TSSK2; TSSK-2
Common Name TSSK2
Gene Symbol TSSK2
Kinase Family G11 Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody)
Kinase Group Ser/Thr Kinases: Atypical
Shipping Condition Dry Ice
System Compatibility Zin-LYTE™
Target Entry STK22B
Target Gene testis-specific serine kinase 2
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length, Recombinant
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.