Learn More
Eurofins Discoverx STK25, active, 10 µg. (14-929)
Supplier:  Eurofins Discoverx 14929
Encompass
N-terminal GST-tagged and C-terminal 6xHis-tagged recombinant, human STK25 amino acids 1-308. STK25 is a member of the germinal centre kinase III (GCK III) subfamily of the sterile 20 superfamily of kinases. The enzyme plays a role in serine-threonine liver kinase B1 (LKB1) signaling pathway to regulate neuronal polarization and morphology of the Golgi apparatus. The protein is translocated from the Golgi apparatus to the nucleus in response to chemical anoxia and plays a role in regulation of cell death. A pseudogene associated with this gene is located on chromosome 18. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9431