Learn More
Aviva Systems Biology Corp Stk25 Antibody - C-terminal region (ARP48703_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48703_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Stk25. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: MGC94619, Stk25ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-285-376