Learn More
Sino Biological Recombinant Human Src Protein (Full Length, GST Tag), Active
Catalog No. 502573121
Encompass_Preferred
Recombinant full length human SRC was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag
Catalog No. 50-257-3121 Supplier Sino Biological Supplier No. S1918G50
Recombinant full length human SRC was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag