Learn More
Aviva Systems Biology Corp STK3 Antibody - N-terminal region (ARP48700_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48700_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against STK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STK3. Alternate names: FLJ90748, KRS1, MST2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59889