Learn More
Aviva Systems Biology Corp STK32A Antibody - N-terminal region (ARP52784_P050) | 100uL
SDP
Catalog No. 5001460830
Encompass_Preferred
This is a rabbit polyclonal antibody against STK32A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STK32A. Alternate names: MGC22688, YANK1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60830 Supplier Aviva Systems Biology Corp Supplier No. ARP52784_P050
This is a rabbit polyclonal antibody against STK32A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human STK32A. Alternate names: MGC22688, YANK1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat