Learn More
Aviva Systems Biology Corp Stk32b Antibody - N-terminal region (ARP52785_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52785_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Stk32b. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2510009F08Rik, STKG6, Stk32, YANK2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-764