Learn More
ABclonal Technology STK38L Rabbit pAb
SDP
Catalog No. 5000107627
Encompass_Preferred
Enables ATP binding activity, magnesium ion binding activity, and protein serine/threonine kinase activity. Involved in intracellular signal transduction. Acts upstream of or within protein phosphorylation. Located in cytosol.
Catalog No. 50-001-07627 Supplier ABclonal Technology Supplier No. A15406500UL
Enables ATP binding activity, magnesium ion binding activity, and protein serine/threonine kinase activity. Involved in intracellular signal transduction. Acts upstream of or within protein phosphorylation. Located in cytosol.