Learn More
Eurofins Discoverx STK39, active, 10 µg. (16-049)
Supplier:  Eurofins Discoverx 16049
Encompass
N-terminal GST-tagged recombinant, human STK39 full length and N-terminal GST-tagged recombinant, human MO25α full length. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9524