Learn More
MSC Emuge-Franken Plug Thread Go/No Go Gage, Steel, #6-48, 2B, 1 ea
Catalog No. 5000515125
Encompass
These plug thread Go/No Go gages are precision tools designed for inspecting the pitch diameter and functional thread of internal-thread components. Manufactured from hardened tool steel, these double-ended gages provide reliable measurement according to ASME B1.2 standards.
  • Designed to inspect pitch diameter and functional thread
  • Constructed from durable hardened tool steel
  • Includes knurled handle for maximum gripping ability and safety
  • Features recessed minor thread diameter for safe functionality
  • Includes reduced thread start diameter for easy insertion
  • Go member side identified with ample space for custom marking
  • No-Go member side identified with red ring
  • Supplied with certificate of accuracy documentation
Catalog No. 50-005-15125 Supplier MSC Supplier No. 93325710
These plug thread Go/No Go gages are precision tools designed for inspecting the pitch diameter and functional thread of internal-thread components. Manufactured from hardened tool steel, these double-ended gages provide reliable measurement according to ASME B1.2 standards.
  • Designed to inspect pitch diameter and functional thread
  • Constructed from durable hardened tool steel
  • Includes knurled handle for maximum gripping ability and safety
  • Features recessed minor thread diameter for safe functionality
  • Includes reduced thread start diameter for easy insertion
  • Go member side identified with ample space for custom marking
  • No-Go member side identified with red ring
  • Supplied with certificate of accuracy documentation