Learn More
MSC Mitutoyo Gage Block, Rectangle, Steel, 50 mm, Grade 0, 1 ea
Catalog No. 5000514913
Encompass
This rectangle steel gage block is a precision measurement tool designed for calibration and layout applications. It features a nominal length of 50 mm and is manufactured to meet Grade 0 accuracy standards, ensuring reliable performance in metalworking and inspection environments.
  • Manufactured from high-quality steel
  • Rectangle shape
  • Grade 0 accuracy
  • Nominal length of 50 mm
  • Compliant with ASME standards
  • Includes certificate of inspection
Catalog No. 50-005-14913 Supplier MSC Supplier No. 07097363
This rectangle steel gage block is a precision measurement tool designed for calibration and layout applications. It features a nominal length of 50 mm and is manufactured to meet Grade 0 accuracy standards, ensuring reliable performance in metalworking and inspection environments.
  • Manufactured from high-quality steel
  • Rectangle shape
  • Grade 0 accuracy
  • Nominal length of 50 mm
  • Compliant with ASME standards
  • Includes certificate of inspection