Learn More
MSC Mitutoyo Square Gage Block Set, Steel, Grade AS-2, 36/Set
Supplier:  MSC 98269475
Encompass
This 36-piece square gage block set consists of high-quality steel blocks designed for precise dimensional measurements and calibration. Each block is manufactured to meet ASME B89.1.9 standards, ensuring high hardness, excellent wear-resistance, and dimensional stability. The set features a square base design for accurate wringing and is supplied in a solid wooden case for protection and organization.
  • Features 36 pieces in total
  • Grade AS-2 accuracy
  • Laser-etched black characters for clear visibility
  • Square base for superior wringing performance
  • Includes Mitutoyo certificate of inspection
  • Constructed from steel for durability and longevity
  • Traceability provided via identification etched into each block
Catalog No. 50-005-12848