Learn More
MSC Bon Tool Co. Wheelbarrow, 6 cu ft, 1 ea
Catalog No. 502816482
Encompass
This steel tray wheelbarrow is designed for heavy-duty use, featuring an 18 gauge steel tray and a robust wheel assembly for durability and ease of transport.
  • Deep, narrow (26 in) 18 gauge steel tray with rolled edges for maximum strength
  • Tire equipped with zerk fitting and ball bearings for smooth operation and easy maintenance
  • Leg stabilizer provides enhanced stability
  • Ships unassembled for convenient delivery
Catalog No. 50-281-6482 Supplier MSC Supplier No. 14058325
This steel tray wheelbarrow is designed for heavy-duty use, featuring an 18 gauge steel tray and a robust wheel assembly for durability and ease of transport.
  • Deep, narrow (26 in) 18 gauge steel tray with rolled edges for maximum strength
  • Tire equipped with zerk fitting and ball bearings for smooth operation and easy maintenance
  • Leg stabilizer provides enhanced stability
  • Ships unassembled for convenient delivery