Learn More
TARGETMOL CHEMICALS INC STL127705 5MG
SDP
Catalog No. 502581388
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. STL127705 (Compound L) is a potent Ku 70/80 heterodimer protein inhibitor and inhibits Ku70/80-DNA interaction with IC50 of 3.5 uM. STL127705 inhibits the activation of Ku-dependent DNA-PKCS kinase with IC50 of 2.5 uM. purity: 99%
Catalog No. 50-258-1388 Supplier TARGETMOL CHEMICALS INC Supplier No. T130175MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. STL127705 (Compound L) is a potent Ku 70/80 heterodimer protein inhibitor and inhibits Ku70/80-DNA interaction with IC50 of 3.5 uM. STL127705 inhibits the activation of Ku-dependent DNA-PKCS kinase with IC50 of 2.5 uM. purity: 99%