Learn More
Medchemexpress LLC HY-122727 5mg Medchemexpress, STL127705 CAS:1326852-06-5 Purity:>98%
SDP
Catalog No. 501969639
Encompass_Preferred
Medchemexpress, HY-122727 5mg STL127705 CAS:1326852-06-5 STL127705 (Compound L) is a Ku 70/80 heterodimer protein inhibitor, inhibits Ku70/80-DNA interaction, with an IC50 of 3.5 μM. STL127705 also inhibits Ku-dependent activation of DNA-PKCS kinase (IC50, 2.5 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9639 Supplier Medchemexpress LLC Supplier No. HY1227275MG
Medchemexpress, HY-122727 5mg STL127705 CAS:1326852-06-5 STL127705 (Compound L) is a Ku 70/80 heterodimer protein inhibitor, inhibits Ku70/80-DNA interaction, with an IC50 of 3.5 μM. STL127705 also inhibits Ku-dependent activation of DNA-PKCS kinase (IC50, 2.5 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.