Learn More
Medchemexpress LLC Stl127705 (compound L) | 1326852-06-5 | 99.0% | 25 MG
SDP
Catalog No. 5000231017
Encompass_Preferred
STL127705 is a small molecule inhibitor of the Ku70/80 heterodimer and DNA-dependent protein kinase (DNA-PK) used in biochemical and cellular studies of DNA repair and apoptosis.
  • Potent Ku70/80 and DNA-PK inhibitor (IC50 3.5 μM and 2.5 μM, respectively).
  • Antiproliferative and apoptosis-inducing activity in cellular assays.
  • High purity (98.97%, HPLC) suitable for research applications.
  • Molecular weight 437.42, formula C22H20FN5O4.
  • Poor aqueous solubility; soluble in DMSO (<1 mg/mL).
  • Stable as powder at -20°C for up to 3 years.
Catalog No. 50-002-31017 Supplier Medchemexpress LLC Supplier No. HY12272725MG
STL127705 is a small molecule inhibitor of the Ku70/80 heterodimer and DNA-dependent protein kinase (DNA-PK) used in biochemical and cellular studies of DNA repair and apoptosis.
  • Potent Ku70/80 and DNA-PK inhibitor (IC50 3.5 μM and 2.5 μM, respectively).
  • Antiproliferative and apoptosis-inducing activity in cellular assays.
  • High purity (98.97%, HPLC) suitable for research applications.
  • Molecular weight 437.42, formula C22H20FN5O4.
  • Poor aqueous solubility; soluble in DMSO (<1 mg/mL).
  • Stable as powder at -20°C for up to 3 years.