Learn More
Cayman Chemical Stm Cel Smal Molcule Scre 50uL
SDP
Catalog No. 502893457
Encompass_Preferred
An Asc-1 inhibitor selectively inhibits Asc-1 in HEK293 cells expressing Asc-1 (IC50 36.8 nM) over 40 other transporters (IC50s >10 UM) inhibits serine cysteine glycine and alanine import and cAMP-induced increases in the OCR and decreases UCP1 levels in primary human white and brown adipose cells at 100 nM decreases the number of malformed neurons and reduces contextual fear memory deficits in a mouse model of surgery-induced brain damage when intracerebroventricularly infused
Catalog No. 50-289-3457 Supplier Cayman Chemical Supplier No. 900182750 UL
An Asc-1 inhibitor selectively inhibits Asc-1 in HEK293 cells expressing Asc-1 (IC50 36.8 nM) over 40 other transporters (IC50s >10 UM) inhibits serine cysteine glycine and alanine import and cAMP-induced increases in the OCR and decreases UCP1 levels in primary human white and brown adipose cells at 100 nM decreases the number of malformed neurons and reduces contextual fear memory deficits in a mouse model of surgery-induced brain damage when intracerebroventricularly infused