Learn More
Lifeline Cell Technology STM CELLS MSC HU AD LIFELN VL
SDP
Catalog No. 50311866
Encompass_Preferred
HMSC-Ad can be expanded in an undifferentiated state for future differentiation to multiple lineages. Lifeline's HMSC-Ad may be differentiated down the typical mesenchymal lineages, such as adipogenic, chondrogenic and osteogenic lineages.
Catalog No. 50-311-866 Supplier Lifeline Cell Technology Supplier No. FC0034
HMSC-Ad can be expanded in an undifferentiated state for future differentiation to multiple lineages. Lifeline's HMSC-Ad may be differentiated down the typical mesenchymal lineages, such as adipogenic, chondrogenic and osteogenic lineages.