Learn More
Lifeline Cell Technology STM CELLS MSC HU LIFELNE VL
SDP
Catalog No. 50311857
Encompass_Preferred
HMSC-WJ and HMSC-Ad can be expanded in an undifferentiated state for future differentiation to multiple lineages. May be differentiated down the typical mesenchymal lineages, such as adipogenic, chondrogenic, and osteogenic lineages.
Catalog No. 50-311-857 Supplier Lifeline Cell Technology Supplier No. FC0020
HMSC-WJ and HMSC-Ad can be expanded in an undifferentiated state for future differentiation to multiple lineages. May be differentiated down the typical mesenchymal lineages, such as adipogenic, chondrogenic, and osteogenic lineages.