Learn More
Lifeline Cell Technology STM CELLS P-ADIP HU LIFELN VL
SDP
Catalog No. 50311882
Encompass_Preferred
Human Pre-Adipocytes can be expanded in an undifferentiated state for future differentiation to mature Adipocytes. Lifeline's Human Pre-Adipocytes may also be differentiated down chondrogenic, and osteogenic lineages.
Catalog No. 50-311-882 Supplier Lifeline Cell Technology Supplier No. FC0062
Human Pre-Adipocytes can be expanded in an undifferentiated state for future differentiation to mature Adipocytes. Lifeline's Human Pre-Adipocytes may also be differentiated down chondrogenic, and osteogenic lineages.