Learn More
Medchemexpress LLC HY-134836 10mg , STM2457 CAS:2499663-01-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13483610MG
Encompass_Preferred
HY-134836 10mg STM2457 CAS: 2499663-01-1 STM2457 is a first-in-class, highly potent, selective and orally active METTL3 inhibitor with an IC50 of 16.9 nM. STM2457 can be used for the research of acute myeloid leukaemia (AML). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9024