Learn More
TARGETMOL CHEMICALS INC STM2457 5MG
SDP
Catalog No. 502579026
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. STM2457 is a first-in-class highly potent selective and orally active inhibitor of METTL3 . It can be used for the research of acute myeloid leukaemia (AML). purity: 99%
Catalog No. 50-257-9026 Supplier TARGETMOL CHEMICALS INC Supplier No. T90605MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. STM2457 is a first-in-class highly potent selective and orally active inhibitor of METTL3 . It can be used for the research of acute myeloid leukaemia (AML). purity: 99%