Learn More
Newcomer Supply Trichrome, Masson, Aniline Blue Stain Kit, 250 ml kit
SDP
Catalog No. NC0827161
Encompass
Newcomer Supply Trichrome, Masson, Aniline Blue Stain Kit procedure, with included microwave modification, is used to differentially demonstrate connective tissue elements, collagen and muscle fibers.
Catalog No. NC0827161 Supplier Newcomer Supply Supplier No. 9179A
Newcomer Supply Trichrome, Masson, Aniline Blue Stain Kit procedure, with included microwave modification, is used to differentially demonstrate connective tissue elements, collagen and muscle fibers.