Learn More
Yamato Scientific America, Inc. Yamato 47L Steam Sterilizer, Standard, 115V
Catalog No. 501898066 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
High performance, easy operation and ergonomically designed 47L standard steam sterilizer featuring easy top loading and handling of samples. Maximum sterilizing temperature goes up to 135°C suitable for protein modification. By simple setting and operation, this sterilizer manages ordinary sterilization, sterilization of culture mediums and liquids, and melting of culture mediums. Custom programs can be saved for repeated use. Standard equipped with timer start and pre-heating features for efficient use of time and cooling fan to shorten cool down time. Mobile on wheels. Operating temp. range 45 to 135°C.
Catalog No. 50-189-8066 Supplier Yamato Scientific America, Inc. Supplier No. SN500C
Add to Cart
Add to Cart
Request a Quote
High performance, easy operation and ergonomically designed 47L standard steam sterilizer featuring easy top loading and handling of samples. Maximum sterilizing temperature goes up to 135°C suitable for protein modification. By simple setting and operation, this sterilizer manages ordinary sterilization, sterilization of culture mediums and liquids, and melting of culture mediums. Custom programs can be saved for repeated use. Standard equipped with timer start and pre-heating features for efficient use of time and cooling fan to shorten cool down time. Mobile on wheels. Operating temp. range 45 to 135°C.