Learn More
Braintree Scientific Std Pattern Dressing Forceps, Serrated Handle, 4.5"" premium
SDP
Catalog No. NC9533940
Encompass
Standard Pattern Dressing Forceps. Premium grade dressing forceps have serrated tips and ribbed grip to assist with delicate dissection - standard pattern, serrated handle. **Sold for research purposes only and not for human use**
Catalog No. NC9533940 Supplier Braintree Scientific Supplier No. FC1001
Standard Pattern Dressing Forceps. Premium grade dressing forceps have serrated tips and ribbed grip to assist with delicate dissection - standard pattern, serrated handle. **Sold for research purposes only and not for human use**