Learn More
Cayman Chemical STng AQ varInthumnreco 100ug
SDP
Catalog No. 502995327
Encompass_Preferred
Source: N-terminal His-tagged human recombinant STING R232, G230A, R293Q mutant purified from E. coli • MW = 28.8 kDa
Catalog No. 50-299-5327 Supplier Cayman Chemical Supplier No. 23592100 UG
Source: N-terminal His-tagged human recombinant STING R232, G230A, R293Q mutant purified from E. coli • MW = 28.8 kDa