Learn More
Cayman Chemical STng H232 varInthumnre 100ug
SDP
Catalog No. 502964354
Encompass_Preferred
Source: N-terminal His-tagged STING catalytic domain purified from E. coli • MW: ~28.8 kDa
Catalog No. 50-296-4354 Supplier Cayman Chemical Supplier No. 22815100 UG
Source: N-terminal His-tagged STING catalytic domain purified from E. coli • MW: ~28.8 kDa