Learn More
Cayman Chemical STng M284 varInt PolycLNal 1ea
SDP
Catalog No. 502934687
Encompass_Preferred
A selective estrogen receptor modulator binds to ERa and ERB (Kis 380 and 410 nM respectively) increases vaginal weight and vaginal epithelial height (ED50s 0.48 and 0.39 mg/kg per day respectively) in an ovariectomized rat model of menopause increases progesterone receptor protein expression in vaginal epithelium and stroma and inhibits 17b-estradiol-induced estrogen response element-mediated transactivation in a reporter assay using MCF-7 cells
Catalog No. 50-293-4687 Supplier Cayman Chemical Supplier No. 247911 EA
A selective estrogen receptor modulator binds to ERa and ERB (Kis 380 and 410 nM respectively) increases vaginal weight and vaginal epithelial height (ED50s 0.48 and 0.39 mg/kg per day respectively) in an ovariectomized rat model of menopause increases progesterone receptor protein expression in vaginal epithelium and stroma and inhibits 17b-estradiol-induced estrogen response element-mediated transactivation in a reporter assay using MCF-7 cells