Learn More
Cayman Chemical STng PROTAC SP23 1mg
SDP
Catalog No. 502918629
Encompass_Preferred
A PROTAC that drives STING degradation; degrades STING in THP-1 cells (DC50 = 3.2 µM); decreases cGAMP-induced increases in the levels of IFN-β, IL-6, and CXCL10 in THP-1 cells at 2.5-10 µM; reduces kidney weight, improves kidney function, and increases the survival rate in a mouse model of cisplatin-induced acute kidney injury at 30 and 60 mg/kg
Catalog No. 50-291-8629 Supplier Cayman Chemical Supplier No. 392741 MG
A PROTAC that drives STING degradation; degrades STING in THP-1 cells (DC50 = 3.2 µM); decreases cGAMP-induced increases in the levels of IFN-β, IL-6, and CXCL10 in THP-1 cells at 2.5-10 µM; reduces kidney weight, improves kidney function, and increases the survival rate in a mouse model of cisplatin-induced acute kidney injury at 30 and 60 mg/kg