Learn More
Cayman Chemical STng R224 varInthumnre 100ug
SDP
Catalog No. 502994735
Encompass_Preferred
Source: N-terminal His-tagged human STING R232 mutant from E. coli • MW: 28.8 kDa
Catalog No. 50-299-4735 Supplier Cayman Chemical Supplier No. 23593100 UG
Source: N-terminal His-tagged human STING R232 mutant from E. coli • MW: 28.8 kDa