Learn More
Electron Microscopy Sciences Stainless Steel Autopsy Cassette
SDP
Catalog No. 5024588
Encompass_Preferred
Made from stainless steel. It is fully perforated allowing for free movement of fluids. It comes with a snap-cap which provides positive enclosure.
Catalog No. 50-245-88 Supplier Electron Microscopy Sciences Supplier No. 6231012
Made from stainless steel. It is fully perforated allowing for free movement of fluids. It comes with a snap-cap which provides positive enclosure.