Learn More
MSC Precision Brand Feeler Gage, 0.020 in., 1/2 in. x 5 in., 10/Pk.
Catalog No. 5000513717
Encompass
This stainless steel thickness gage is designed for precise measurement applications, featuring a 0.508 mm (0.020 in.) thickness and a durable 300 series stainless steel construction. It is engineered for reliability in diverse industrial settings.
  • Constructed from 300 series stainless steel for corrosion resistance
  • Low magnetic properties suitable for sensitive environments
  • Withstands high temperatures
  • Includes 10 pieces per package
  • Features thickness etched on the material for easy identification
  • Ideal for use in aircraft, power/utility, food, automotive, and marine industries
  • Hardness rating of 45-55 Rockwell Hardness C scale
Catalog No. 50-005-13717 Supplier MSC Supplier No. 67822304
This stainless steel thickness gage is designed for precise measurement applications, featuring a 0.508 mm (0.020 in.) thickness and a durable 300 series stainless steel construction. It is engineered for reliability in diverse industrial settings.
  • Constructed from 300 series stainless steel for corrosion resistance
  • Low magnetic properties suitable for sensitive environments
  • Withstands high temperatures
  • Includes 10 pieces per package
  • Features thickness etched on the material for easy identification
  • Ideal for use in aircraft, power/utility, food, automotive, and marine industries
  • Hardness rating of 45-55 Rockwell Hardness C scale