Learn More
TARGETMOL CHEMICALS INC STO-609 10MG
SDP
Catalog No. 502876194
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. STO-609 is a specific and cell-permeable inhibitor of the Ca2+/calmodulin-dependent protein kinase kinase (CaM-KK) for recombinant CaM-KKalpha/KKbeta (Ki 80/15 ng/mL). Purity 98.95%
Catalog No. 50-287-6194 Supplier TARGETMOL CHEMICALS INC Supplier No. T354610MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. STO-609 is a specific and cell-permeable inhibitor of the Ca2+/calmodulin-dependent protein kinase kinase (CaM-KK) for recombinant CaM-KKalpha/KKbeta (Ki 80/15 ng/mL). Purity 98.95%