Learn More
Selleck Chemical LLC STO-609
SDP
Supplier:  Selleck Chemical LLC S8274100MG
Encompass_Preferred
STO-609 is a specific inhibitor of the Ca2 /Calmodulin-dependent protein kinase kinase(CaM-KK) that inhibits the activities of recombinant CaM-KK and CaM-KK isoforms with Ki values of 80 and 15 ng/ml respectively and also inhibits their autophosphorylation activities STO-609 inhibits AMPKK activity and inhibits autophagy
Catalog No. 50-000-00930