Learn More
Medchemexpress LLC STO-609 | 52029-86-4 | 98.2% | 314.29 | 100 MG
SDP
Catalog No. 5000402064
Encompass_Preferred
STO-609 is a selective and cell-permeable inhibitor of the Ca2+/calmodulin-dependent protein kinase kinase (CaM-KK), with Ki values of 80 and 15 ng/mL for recombinant CaM-KKα and CaM-KKβ, respectively. It inhibits AMP-activated protein kinase kinase (AMPKK) activity in HeLa cell lysates with an IC50 ~0.02 g/ml.
  • Selective inhibitor of CaM-KK
  • Cell-permeable
  • Inhibits AMPKK activity
Catalog No. 50-004-02064 Supplier Medchemexpress LLC Supplier No. HY19805100MG
STO-609 is a selective and cell-permeable inhibitor of the Ca2+/calmodulin-dependent protein kinase kinase (CaM-KK), with Ki values of 80 and 15 ng/mL for recombinant CaM-KKα and CaM-KKβ, respectively. It inhibits AMP-activated protein kinase kinase (AMPKK) activity in HeLa cell lysates with an IC50 ~0.02 g/ml.
  • Selective inhibitor of CaM-KK
  • Cell-permeable
  • Inhibits AMPKK activity