Learn More
Medchemexpress LLC Stobadine | 85202-17-1 | 99.4% | 202.30 g/mol | C13H18N2 | 1 MG
SDP
Catalog No. 5000210522
Encompass_Preferred
Stobadine is a small-molecule antioxidant used in research to prevent free-radical-induced alterations in membrane fluidity and to study oxidative stress-related mechanisms in biochemical and pharmacological assays.
  • Antioxidant and free radical scavenger.
  • Used in studies of membrane fluidity and cardioprotective mechanisms.
  • White to off-white solid suitable for preparing solutions for assays.
  • High reported purity for reliable experimental results.
  • Recommended storage at low temperatures to preserve stability.
Catalog No. 50-002-10522 Supplier Medchemexpress LLC Supplier No. HYW0675721MG
Stobadine is a small-molecule antioxidant used in research to prevent free-radical-induced alterations in membrane fluidity and to study oxidative stress-related mechanisms in biochemical and pharmacological assays.
  • Antioxidant and free radical scavenger.
  • Used in studies of membrane fluidity and cardioprotective mechanisms.
  • White to off-white solid suitable for preparing solutions for assays.
  • High reported purity for reliable experimental results.
  • Recommended storage at low temperatures to preserve stability.