Learn More
Grainger Durham Manufacturing Mesh Stock Truck, 2 Shelves, 2,000 lb Capacity, 1 ea
Catalog No. 502791655
Encompass_Preferred
This two-sided mesh stock truck is designed for efficient material handling, featuring a durable construction with two fixed shelves. The unit is equipped with heavy-duty casters to provide mobility for loads up to 2,000 lbs.
  • Features a tubular push handle for easy maneuvering.
  • Includes two rigid and two swivel 6 in. x 2 in. phenolic casters.
  • Constructed with a 14 gauge mesh design and a textured powder coat finish.
  • Provides two fixed shelves with a flush lip design.
  • Overall dimensions are 30-1/4 in. W x 54-1/2 in. D x 56-7/16 in. H.
  • Includes a 22 in. clearance between shelves.
Catalog No. 50-279-1655 Supplier Grainger Supplier No. 16C313
This two-sided mesh stock truck is designed for efficient material handling, featuring a durable construction with two fixed shelves. The unit is equipped with heavy-duty casters to provide mobility for loads up to 2,000 lbs.
  • Features a tubular push handle for easy maneuvering.
  • Includes two rigid and two swivel 6 in. x 2 in. phenolic casters.
  • Constructed with a 14 gauge mesh design and a textured powder coat finish.
  • Provides two fixed shelves with a flush lip design.
  • Overall dimensions are 30-1/4 in. W x 54-1/2 in. D x 56-7/16 in. H.
  • Includes a 22 in. clearance between shelves.