Learn More
Inorganic Ventures ICP TUNING SOLUTION 24 125mL
SDP
Catalog No. 507527032
Encompass_Preferred
Inorganic Ventures' Stock Tuning Solution 24 is a 500mL certified reference material set in a nitric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-752-7032 Supplier Inorganic Ventures Supplier No. IVSTOCK24125ML
Inorganic Ventures' Stock Tuning Solution 24 is a 500mL certified reference material set in a nitric acid matrix for stability. This standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search