Learn More
Med Vet International McKesson Sterile Impervious Surgical Stockinette, 9 W x 48 L Inch, 16-3M-948, Each
Catalog No. 503044055
Encompass_Preferred
Surgical Stockinette Impervious / Tubular McKesson 9 W X 48 L Inch Sterile 9 x 48 Inches Textured polyester knit liner with impervious tube made of elastomeric film Precut in convenient sizes, sewn, rolled, and packaged in CSR (central supply room) wrap
Catalog No. 50-304-4055 Supplier Med Vet International Supplier No. 460836EA
Surgical Stockinette Impervious / Tubular McKesson 9 W X 48 L Inch Sterile 9 x 48 Inches Textured polyester knit liner with impervious tube made of elastomeric film Precut in convenient sizes, sewn, rolled, and packaged in CSR (central supply room) wrap