Learn More
Med Vet International Stockinette Tubular / Undercast 3M Synthetic 2 inch X 25 Yard Polyester Nonsterile, Ms02, 1 Roll
Catalog No. 502829666
Encompass_Preferred
Water repellent and therefore suitable for use with Soft Cast as a waterproof cast Suitable for use with all casting materials, plaster and synthetic 2 inches x 25 yards
Catalog No. 50-282-9666 Supplier Med Vet International Supplier No. 131514CS
Water repellent and therefore suitable for use with Soft Cast as a waterproof cast Suitable for use with all casting materials, plaster and synthetic 2 inches x 25 yards