Learn More
Med Vet International Stockinette Tubular / Undercast 3M Synthetic 4 inch X 25 Yard Polyester Nonsterile, Ms04, 1 Roll
Catalog No. 502831241
Encompass_Preferred
Water repellent and therefore suitable for use with Soft Cast as a waterproof cast Suitable for use with all casting materials, plaster and synthetic 4 inches x 25 yards
Catalog No. 50-283-1241 Supplier Med Vet International Supplier No. 131516CS
Water repellent and therefore suitable for use with Soft Cast as a waterproof cast Suitable for use with all casting materials, plaster and synthetic 4 inches x 25 yards