Learn More
Med Vet International Stockinette Tubular / Undercast 3M Synthetic 6 inch X 25 Yard Polyester Nonsterile, Ms06, 1 Roll
Catalog No. 502831780
Encompass_Preferred
Water repellent and therefore suitable for use with Soft Cast as a waterproof cast Suitable for use with all casting materials, plaster and synthetic Not made with natural rubber latex
Catalog No. 50-283-1780 Supplier Med Vet International Supplier No. 199246CS
Water repellent and therefore suitable for use with Soft Cast as a waterproof cast Suitable for use with all casting materials, plaster and synthetic Not made with natural rubber latex