Learn More
Med Vet International McKesson Beige Cotton Tubular Stockinette, 4 x 48 Inch, 16-3S-448, Each
Catalog No. 503043522
Encompass_Preferred
Surgical Stockinette Tubular McKesson 4 X 48 Inch Sterile Knitted from high-grade yarn and inspected to guarantee consistent knit construction, cleanliness, and proper roll dimensions Provides pressure to hold dressing in place Packaged in an easy-to-use dispenser box on a continuous roll, allowing user to determine length Cotton Beige 4 x 48 inches
Catalog No. 50-304-3522 Supplier Med Vet International Supplier No. 457658EA
Surgical Stockinette Tubular McKesson 4 X 48 Inch Sterile Knitted from high-grade yarn and inspected to guarantee consistent knit construction, cleanliness, and proper roll dimensions Provides pressure to hold dressing in place Packaged in an easy-to-use dispenser box on a continuous roll, allowing user to determine length Cotton Beige 4 x 48 inches