Learn More
Med Vet International McKesson Beige Cotton Tubular Stockinette, 2 Inch x 25 Yard, 16-4T-225, Each
Catalog No. 503043423
Encompass_Preferred
Stockinette Tubular McKesson 2 Inch X 25 Yard Cotton NonSterile Knitted from high-grade yarn and inspected to guarantee consistent knit construction, cleanliness, and proper roll dimensions Provides pressure to hold dressing in place Packaged in an easy-to-use dispenser box on a continuous roll, allowing user to determine length Cotton Beige 2 inches x 25 yards
Catalog No. 50-304-3423 Supplier Med Vet International Supplier No. 457629RL
Stockinette Tubular McKesson 2 Inch X 25 Yard Cotton NonSterile Knitted from high-grade yarn and inspected to guarantee consistent knit construction, cleanliness, and proper roll dimensions Provides pressure to hold dressing in place Packaged in an easy-to-use dispenser box on a continuous roll, allowing user to determine length Cotton Beige 2 inches x 25 yards